Orders LinkContactUsLinkComponent
Cesio cloruro
Cesio cloruro
Glass bottle for solids, 1 kg
Catalog # A1098.1000
Supplier:   PanReac AppliChem
Cesio cloruro
Catalog # A1098.1000
Supplier:  PanReac AppliChem
CAS Number:  7647-17-8
Cesio cloruro
Glass bottle for solids, 1 kg
CAS Number:  7647-17-8
Price
SFr. 1’700.00
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  Cesium Chloride
  • CAS Number:7647-17-8
  • Formula:CsCl
  • MDL Number:MFCD00010955
  • Molecular Weight:168.36 g/mol
  • Merck Index:13,02021
  • Boiling Pt:1303 °C (1013 hPa)
  • Melting Pt:646 °C
  • Density:3,7808 g/cm³ (20 °C)
  • Storage Temperature:Ambiente
  • EINECS:231-600-2
Specifications
Specifications
  • Tipo di conf.:
    Glass bottle for solids
  • Conf:
    1 kg