Microplates, Well-Coated™
Informazioni su questo articolo
Pre-coated, blocked microtitre plates for screening and rapid assays.
- Available in clear, black or white for colorimetric, chemiluminescent and fluorescent detection systems
- 8 well strip format, supplied in convenient 96 well holder
- Pre-blocked to save time
Protein A, G or A/G
Binds the constant (Fc) region of immunoglobulins ensuring that the antigen binding domain of the antibody is orientated away from the plate, offering maximum exposure of the binding site.
Protein L
Binds the kappa light chains of immunoglobulins without interfering with the antigen binding site; binds to all classes of immunoglobulin, including IgG, IgM, IgA, IgE and IgD. Also binds to single chain variable fragments (scFv and Fab).
Goat α-mouse IgG antibody or α-rabbit IgG antibody
Suitable for binding assays using low quantities of antibodies or antibodies that denature on direct binding to polystyrene plates.
Biotin binding
Designed to specifically bind avidin, steptavidin or Neutravidin™ conjugated molecules, including enzyme conjugates.
Nickel
Binding of 6X His tagged, expressed recombinant proteins.
Glutathione
Specific binding of GST tagged, expressed recombinant proteins.
Amine or sulfhydryl binding
Custom design ELISA plates and couple amine or sulfhydryl containing peptides, proteins, and compounds.
39 varianti
Product Details & Documents
Pre-coated, blocked microtitre plates for screening and rapid assays.
- Available in clear, black or white for colorimetric, chemiluminescent and fluorescent detection systems
- 8 well strip format, supplied in convenient 96 well holder
- Pre-blocked to save time
Protein A, G or A/G
Binds the constant (Fc) region of immunoglobulins ensuring that the antigen binding domain of the antibody is orientated away from the plate, offering maximum exposure of the binding site.
Protein L
Binds the kappa light chains of immunoglobulins without interfering with the antigen binding site; binds to all classes of immunoglobulin, including IgG, IgM, IgA, IgE and IgD. Also binds to single chain variable fragments (scFv and Fab).
Goat α-mouse IgG antibody or α-rabbit IgG antibody
Suitable for binding assays using low quantities of antibodies or antibodies that denature on direct binding to polystyrene plates.
Biotin binding
Designed to specifically bind avidin, steptavidin or Neutravidin™ conjugated molecules, including enzyme conjugates.
Nickel
Binding of 6X His tagged, expressed recombinant proteins.
Glutathione
Specific binding of GST tagged, expressed recombinant proteins.
Amine or sulfhydryl binding
Custom design ELISA plates and couple amine or sulfhydryl containing peptides, proteins, and compounds.
Documents
Product Family Options
Protein A, G or A/G
Informazioni sul prodotto
- ColoreDisponibilitàPrice
- 786-731Well-Coated™ Protein A 8-well strip plateSpecifiche:
More
Specifications
Cat. No.786-731 - 786-770Well-Coated™ Protein A 96-well plateSpecifiche:
More
Specifications
Cat. No.786-770 - 786-771Well-Coated™ Protein A 96-well plateSpecifiche:
More
Specifications
Cat. No.786-771 - 786-733Well-Coated™ Protein G 8-well strip plateSpecifiche:
More
Specifications
Cat. No.786-733 - 786-774Well-Coated™ Protein G 96-well plateSpecifiche:
More
Specifications
Cat. No.786-774 - 786-775Well-Coated™ Protein G 96-well plateSpecifiche:
More
Specifications
Cat. No.786-775 - 786-735Well-Coated™ Protein A/G 8-well strip plateSpecifiche:
More
Specifications
Cat. No.786-735 - 786-772Well-Coated™ Protein A/G 96-well plateSpecifiche:
More
Specifications
Cat. No.786-772 - 786-773Well-Coated™ Protein A/G 96-well plateSpecifiche:
More
Specifications
Cat. No.786-773
Protein L
Informazioni sul prodotto
- ColoreDisponibilitàPrice
- 786-737Well-Coated™ Protein L 8-well strip plateSpecifiche:
More
Specifications
Cat. No.786-737 - 786-776Well-Coated™ Protein L 96-well plateSpecifiche:
More
Specifications
Cat. No.786-776 - 786-777Well-Coated™ Protein L 96-well plateSpecifiche:
More
Specifications
Cat. No.786-777
Goat α-mouse IgG antibody or α-rabbit IgG antibody
Informazioni sul prodotto
- ColoreDisponibilitàPrice
- 786-739Well-Coated™ Antibody (goat α-mouse) 8-well strip plateWell-Coated™ Antibody (goat α-mouse) 8-well strip platetrasparenteLog in per disponibilità1 * 5 STSFr. 323.00Specifiche:
More
Specifications
Cat. No.786-739 - 786-758Well-Coated™ Antibody (goat α-mouse) 96-well plateSpecifiche:
More
Specifications
Cat. No.786-758 - 786-759Well-Coated™ Antibody (goat α-mouse) 96-well plateWell-Coated™ Antibody (goat α-mouse) 96-well platebiancoLog in per disponibilità1 * 5 STSFr. 305.00Specifiche:
More
Specifications
Cat. No.786-759 - 786-741Well-Coated™ Antibody (goat α-rabbit) 8-well strip plateWell-Coated™ Antibody (goat α-rabbit) 8-well strip platetrasparenteLog in per disponibilità1 * 5 STSFr. 323.00Specifiche:
More
Specifications
Cat. No.786-741 - 786-760Well-Coated™ Antibody (goat α-rabbit) 96-well plateWell-Coated™ Antibody (goat α-rabbit) 96-well plateneroLog in per disponibilità1 * 5 STSFr. 305.00Specifiche:
More
Specifications
Cat. No.786-760 - 786-761Well-Coated™ Antibody (goat α-rabbit) 96-well plateWell-Coated™ Antibody (goat α-rabbit) 96-well platebiancoLog in per disponibilità1 * 5 STSFr. 305.00Specifiche:
More
Specifications
Cat. No.786-761
Biotin binding
Informazioni sul prodotto
- ColoreDisponibilitàPrice
- 786-743Well-Coated™ Neutravidin 8-well strip plateSpecifiche:
More
Specifications
Cat. No.786-743 - 786-766Well-Coated™ Neutravidin 96-well plateSpecifiche:
More
Specifications
Cat. No.786-766 - 786-767Well-Coated™ Neutravidin 96-well plateSpecifiche:
More
Specifications
Cat. No.786-767 - 786-745Well-Coated™ Streptavidin 8-well strip plateWell-Coated™ Streptavidin 8-well strip platetrasparenteLog in per disponibilità1 * 5 STSFr. 331.00Specifiche:
More
Specifications
Cat. No.786-745 - 786-778Well-Coated™ Streptavidin 96-well plateSpecifiche:
More
Specifications
Cat. No.786-778 - 786-779Well-Coated™ Streptavidin 96-well plateSpecifiche:
More
Specifications
Cat. No.786-779 - 786-747Well-Coated™ Biotin 8-well strip plateSpecifiche:
More
Specifications
Cat. No.786-747 - 786-762Well-Coated™ Biotin 96-well plateSpecifiche:
More
Specifications
Cat. No.786-762 - 786-763Well-Coated™ Biotin 96-well plateSpecifiche:
More
Specifications
Cat. No.786-763
Nickel
Informazioni sul prodotto
- ColoreDisponibilitàPrice
- 786-749Well-Coated™ Nickel 8-well strip plateSpecifiche:
More
Specifications
Cat. No.786-749 - 786-768Well-Coated™ Nickel 96-well plateSpecifiche:
More
Specifications
Cat. No.786-768 - 786-769Well-Coated™ Nickel 96-well plateSpecifiche:
More
Specifications
Cat. No.786-769
Glutathione
Informazioni sul prodotto
- ColoreDisponibilitàPrice
- 786-751Well-Coated™ Glutathione 8-well strip plateSpecifiche:
More
Specifications
Cat. No.786-751 - 786-764Well-Coated™ Glutathione 96-well plateSpecifiche:
More
Specifications
Cat. No.786-764 - 786-765Well-Coated™ Glutathione 96-well plateSpecifiche:
More
Specifications
Cat. No.786-765
Amine or sulfhydryl binding
Informazioni sul prodotto
- ColoreDisponibilitàPrice
- 786-753Well-Coated™ Amine binding 8-well strip plateWell-Coated™ Amine binding 8-well strip platetrasparenteLog in per disponibilità1 * 5 STSFr. 238.00Specifiche:
More
Specifications
Cat. No.786-753 - 786-756Well-Coated™ Amine binding 96-well plateSpecifiche:
More
Specifications
Cat. No.786-756 - 786-757Well-Coated™ Amine binding 96-well plateSpecifiche:
More
Specifications
Cat. No.786-757 - 786-755Well-Coated™ Sulfhydryl binding 8-well strip plateWell-Coated™ Sulfhydryl binding 8-well strip platetrasparenteLog in per disponibilità1 * 5 STSFr. 246.00Specifiche:
More
Specifications
Cat. No.786-755 - 786-780Well-Coated™ Sulfhydryl binding 96-well plateSpecifiche:
More
Specifications
Cat. No.786-780 - 786-781Well-Coated™ Sulfhydryl binding 96-well plateSpecifiche:
More
Specifications
Cat. No.786-781