2-Methyl-8-quinolinol
Glass bottle for solids, 100 g
MOLE20572352-100G
Marketsource
: Molekula
: 8-Hydroxy-2-methylquinoline, 8-Hydroxyquinaldine, 2-Methyloxine
- :826-81-3
- :C₁₀H₉NO
- :MFCD00006765
- :159.19 g/mol
- :267 °C (1013 hPa)
- :70…72 °C
- :Ambiente
- :212-562-6
- :3077
- :9,III
- Tipo di conf.:Glass bottle for solids
- Conf:100 g