Stopcocks for burettes, angled, DURAN®
2,0 mm, 12,5 NS
Codice catalogo SCOT286451608
Nome del fornitore: DWK Life Sciences
Stopcocks for burettes, angled, DURAN®
Codice catalogo SCOT286451608
Nome del fornitore: DWK Life Sciences
Vai a
Product Details & Documents
DURAN®, borosilicate glass 3.3, clear, grounded and polished, without key.
- Very good chemical resistance
- Very good temperature resistance
For glass manipulators.
Specifiche
- Ø foro:2,0 mm
- Rubinetto:12,5 NS