Tubing connectors, Magic box, LaboPlast®
229-7420
: BURKLE
- Ø int.tubo:4 - 17 mm
- Conf:1 SET
Magic box contains 68 connectors.
- Good chemical resistance
- Good temperature resistance
In this magic box, you can invariably find the appropriate hose connector you are looking for.
: Includes straight hose connector, T hose connector,Y hose connector, X hose connector, angle hose connector (two of each in sizes 4, 6, 8, 10, 12 and 14 mm), universal hose connector (two of each for hoses from 4 to 17 mm), reducing hose connector (two of each in sizes 4 to 8, 4 to 12, and 8 to 12 mm).