Tubing connectors, complete with screw cap, straight, DURAN®
Catalog # SCOT286221806
Supplier: DWK Life Sciences
Specifications
- Filettatura:GL 14
- Ø int.tubo:8 mm
- Conf:10
Specifications
About this item
Screw cap PTB, red, with white PP straight hose connection.
- Very good chemical resistance
- Very good temperature resistance
With silicone seal (VMQ).