α-Bromo-4-nitrotoluene
Glass bottle for solids, 100 g
MOLE10671675-100G
Marketsource
: Molekula
: 4-Nitrobenzyl bromide, 1-(Bromomethyl)-4-nitrobenzene, α-Bromo-p-nitrotoluene
- :100-11-8
- :C₇H₆BrNO₂
- :MFCD00007373
- :216.03 g/mol
- :304 °C (1013 hPa)
- :94…96 °C
- :Ambiente
- :202-820-6
- :3261
- :8,II
- Tipo di conf.:Glass bottle for solids
- Conf:100 g