Bunsen burners, with stopcock
Catalog # 451-0005
Supplier: BOCHEM
Specifications
- Mass. temp.:1300 °C
- Descrizione:Becco Bunsen multigas, con rubinetto e regolazione dell'aria
- Conf:1
- Per:All gases
Specifications
About this item
This multigas burner can reach a maximum temperature of 1300 °C and has an anti-slip rubber coating under the base.
- With air regulation