Teclu burners, all gases
BOCH7430
: BOCHEM
- Mass. temp.:1300 °C
- Descrizione:Bruciatore multigas, con rubinetto e regolazione dell'aria
- Conf:1
- Per:All gases
Multi gas burner with stopcock works to a maximum of 1300 °C.
- With air regulation
- With anti-slip rubber coating under the base