Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
 

Serotonin

500 µl
Catalog # CHMCAB938
Supplier:   Chemicon
Serotonin
Catalog # CHMCAB938
Supplier:  Chemicon
CAS Number:  50-67-9
Serotonin

500 µl

CAS Number:  50-67-9
Price
SFr. 1’460.00
Log in for availability
1 * 500 µl
Product Details & Documents
  • CAS Number:50-67-9
  • Formula:C₁₀H₁₂N₂O
  • Molecular Weight:176.22 g/mol
  • EINECS:200-058-9
Specifications

Specifications

  • Conf:
    500 µl

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.