ContactUsLinkComponent
Bicchieri graduati, con manico
  BOCH8650
 :  BOCHEM
 : 8650
undefined
Bicchieri graduati, con manico
  BOCH8650
 :  BOCHEM
 : 8650
Select an Option
125 mm
Select an Option
500 ml
Select an Option
95 mm
Select an Option
1
Stainless steel, 18/10, with handle and spout.
  • Good chemical resistance
  • Excellent resistance to heat
  • Smooth surfaces for fast, easy cleaning
Ideal for corrosive materials. Robust and durable.
Frequently Bought Together