Anisolo, Supelco®
: Merck
: Methyl phenyl ether, Methoxybenzene
- :100-66-3
- :C₇H₈O
- :MFCD00008354
- :108.14 g/mol
- :13,00672
- :154 °C (1013 hPa)
- :–37 °C
- :0,9961 g/cm³ (20 °C)
- :44 °C
- :Ambiente
- :2222
- :3,III
- :M-Clarity™ quality level = MQ100
- :202-876-1
: Methyl phenyl ether, Methoxybenzene
- :100-66-3
- :C₇H₈O
- :MFCD00008354
- :108.14 g/mol
- :13,00672
- :154 °C (1013 hPa)
- :–37 °C
- :0,9961 g/cm³ (20 °C)
- :44 °C
- :Ambiente
- :2222
- :3,III
- :M-Clarity™ quality level = MQ100
- :202-876-1
Product Family Options
- Tipo di conf.Conf