Orders LinkContactUsLinkComponent
Burettes, straight stopcock, PYREX®
Burettes, straight stopcock, PYREX®
Supplier:   SciLabware
Burettes, straight stopcock, PYREX®
Supplier:  SciLabware
Certificates
Burettes, straight stopcock, PYREX®
Burettes, straight stopcock, PYREX®
8 Options Available Below
Product Details & Documents
Borosilicate glass with interchangeable PTFE key, Class AS.
  • Calibrated to deliver ‘Ex’
  • PTFE key for grease-free perfomance
  • White enamel graduations for good visibility with dark coloured solutions
Certifications: ISO 385.
Documents
Product Family Options
Class AS burettes
Product Information
  • Capacità
    Divisione
    Tolleranza
    Availability
    Price
  • 10 ml
    0,02 ml
    0,02 ± ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 63.00
    SFr. 63.00
    1 * 1 ST
       SFr. 63.00
  • 25 ml
    0,10 ml
    0,05 ± ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 53.00
    SFr. 53.00
    1 * 1 ST
       SFr. 53.00
  • 50 ml
    0,10 ml
    0,05 ± ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 55.00
    SFr. 55.00
    1 * 1 ST
       SFr. 55.00
  • 100 ml
    0,20 ml
    0,10 ± ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 73.00
    SFr. 73.00
    1 * 1 ST
       SFr. 73.00
Class AS burettes, works certified
Product Information
  • Capacità
    Divisione
    Tolleranza
    Availability
    Price
  • 10 ml
    0,02 ml
    0,02 ± ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 106.00
    SFr. 106.00
    1 * 1 ST
       SFr. 106.00
  • 25 ml
    0,10 ml
    0,05 ± ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 76.30
    SFr. 76.30
    1 * 1 ST
       SFr. 76.30
  • 50 ml
    0,10 ml
    0,05 ± ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 76.30
    SFr. 76.30
    1 * 1 ST
       SFr. 76.30
  • 100 ml
    0,20 ml
    0,10 ± ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 129.00
    SFr. 129.00
    1 * 1 ST
       SFr. 129.00