Orders LinkContactUsLinkComponent
Etichette
Etichette
Supplier:   KARTELL
Etichette
Supplier:  KARTELL
Certificates
Etichette
8 Options Available Below
Product Details & Documents
Transparent, punched labels for use with wash bottles and plastic or glass reagent bottles.
  • Complete with risk assessment numbers
  • Recommended by the NFPA (National Fire Protection Association)
Product Family Options
Product Information
  • Descrizione
    Colore
    Dimensioni
    Availability
    Price
  • Etichette per acetone
    rosso
    130×35 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - SFr. 368.00
    SFr. 368.00
    1 * 100 ST
       SFr. 368.00
  • Etichette per acqua distillata
    blu
    130×35 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - SFr. 539.00
    SFr. 539.00
    1 * 100 ST
       SFr. 539.00
  • Etichette per etanolo
    verde
    130×35 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - SFr. 368.00
    SFr. 368.00
    1 * 100 ST
       SFr. 368.00
  • Etichette per metanolo
    nero
    130×35 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - SFr. 368.00
    SFr. 368.00
    1 * 100 ST
       SFr. 368.00
  • Etichette per isopropanolo
    marrone
    130×35 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - SFr. 368.00
    SFr. 368.00
    1 * 100 ST
       SFr. 368.00
  • Etichette per etilacetato
    arancione
    130×35 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - SFr. 368.00
    SFr. 368.00
    1 * 100 ST
       SFr. 368.00
  • Etichette per cloroformio
    nero
    130×35 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - SFr. 368.00
    SFr. 368.00
    1 * 100 ST
       SFr. 368.00
  • Etichette per diclorometano
    nero
    130×35 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - SFr. 368.00
    SFr. 368.00
    1 * 100 ST
       SFr. 368.00