Cooling Boxes
Nome del fornitore: KARTELL
Informazioni su questo articolo
Cooling boxes are low temperature containers made of high impact polystyrene material, thermally isolated by means of a polyurethane isolating jacket.
- Non-toxic
- 100% food safe
Informazioni per l'ordine: Cooling block is available for cooling boxes.
2 varianti
Product Details & Documents
Cooling boxes are low temperature containers made of high impact polystyrene material, thermally isolated by means of a polyurethane isolating jacket.
- Non-toxic
- 100% food safe
Informazioni per l'ordine: Cooling block is available for cooling boxes.
Product Family Options
Informazioni sul prodotto
Accessories
Informazioni sul prodotto
- Codice catalogoCapacitàL×P×ADisponibilitàPrice
- HDPE cooling block for Kartell Frigo boxHDPE cooling block for Kartell Frigo boxKART3707500 ml17,5×9×3 cmLog in per disponibilità1 * 1 ST - SFr. 10.90