pH combination electrodes, ProcessLine
About this item
These electrodes are low maintenance sensors for heavy duty process applications in the chemical industry. They are ideally suitable for measuring media with extreme ionic strength - whether boiler feed water or brine - also in strongly oxidizing acid and alkali containing media.
- Low-maintenance electrodes: no electrolyte topping up or installation of complicated pressure sequence control required
- Pinhole diaphragms, so reference electrode is not polluted or blocked
- Duralid electrolyte with high KCl content combines very long working life with rapid and stable measurements
- Proven H membrane glass with very low alkali error and optimized spherical shape
- Integrated pressure compensation cushion
PL8× models
Reference system: Duralid solid electrolyte with a high KCl content
Diaphragm: 2-hole
PL9× models
Reference system: RheoLid electrolyte with KCI storage rings
Diaphragm: PTFE
41 Options Available Below
Product Details & Documents
These electrodes are low maintenance sensors for heavy duty process applications in the chemical industry. They are ideally suitable for measuring media with extreme ionic strength - whether boiler feed water or brine - also in strongly oxidizing acid and alkali containing media.
- Low-maintenance electrodes: no electrolyte topping up or installation of complicated pressure sequence control required
- Pinhole diaphragms, so reference electrode is not polluted or blocked
- Duralid electrolyte with high KCl content combines very long working life with rapid and stable measurements
- Proven H membrane glass with very low alkali error and optimized spherical shape
- Integrated pressure compensation cushion
PL8× models
Reference system: Duralid solid electrolyte with a high KCl content
Diaphragm: 2-hole
PL9× models
Reference system: RheoLid electrolyte with KCI storage rings
Diaphragm: PTFE
Documents
SI Analytics Process Equipment Catalog VWR Process Equipment Catalog.pdf
Product Literature
Product Family Options
Product Information
- TipopHTemperaturaMaterialeØ×LConnettoreAvailabilityPrice
- PL 81-120pHT VP, with Pt10000 - 140...+130 °CVetro12×120 mmVP screw plugLog in for availability1 * 1 STSFr. 578.00
- PL 81-225pHT VP, with Pt10000 - 140...+130 °CVetro12×225 mmVP screw plugLog in for availability1 * 1 STSFr. 687.00
- PL 81-325pHT VP, with Pt10000 - 140...+130 °CVetro12×325 mmVP screw plugLog in for availability1 * 1 STSFr. 737.00
- PL 81-360pHT VP, with Pt10000 - 140...+130 °CVetro12×360 mmVP screw plugLog in for availability1 * 1 STSFr. 781.00
- PL 81-425pHT VP, with Pt10000 - 140...+130 °CVetro12×425 mmVP screw plugLog in for availability1 * 1 STSFr. 803.00
- PL 82-120pHT VP, with Pt1000 - 140...+130 °CVetro12×120 mmVP screw plugLog in for availability1 * 1 STSFr. 622.00
- PL 82-225pHT VP, with Pt1000 - 140...+130 °CVetro12×225 mmVP screw plugLog in for availability1 * 1 STSFr. 693.00
- PL 82-325pHT VP, with Pt1000 - 140...+130 °CVetro12×325 mmVP screw plugLog in for availability1 * 1 STSFr. 750.00
- PL 82-360pHT VP, with Pt1000 - 140...+130 °CVetro12×360 mmVP screw plugLog in for availability1 * 1 STSFr. 797.00
- PL 82-425pHT VP, with Pt1000 - 140...+130 °CVetro12×425 mmVP screw plugLog in for availability1 * 1 STSFr. 810.00
- PL 83-225 NMSN, with NTC300 - 140...+130 °CVetro12×225 mmMemosens® screw plugLog in for availability1 * 1 STSFr. 812.00
- PL 83-325 NMSN, with NTC300 - 140...+130 °CVetro12×325 mmMemosens® screw plugLog in for availability1 * 1 STSFr. 866.00
- PL 83-360 NMSN, with NTC300 - 140...+130 °CVetro12×360 mmMemosens® screw plugLog in for availability1 * 1 STSFr. 902.00
- PL 83-425 NMSN, with NTC300 - 140...+130 °CVetro12×425 mmMemosens® screw plugLog in for availability1 * 1 STSFr. 927.00
- PL 89-120Pt, redox0 - 140...+130 °CVetro12×120 mmPg 13.5Log in for availability1 * 1 STSFr. 687.00
- PL 89-120 NMSN, with NTC30—0...+130 °CVetro12×120 mmMemosens® screw plugLog in for availability1 * 1 STSFr. 824.00
- PL 89-225 NMSN, with NTC30—0...+130 °CVetro12×225 mmMemosens® screw plugLog in for availability1 * 1 STSFr. 891.00
- PL-S 82-225pHT VP, with Pt1000 - 140...+130 °CVetro12×225 mmVP screw plugLog in for availability1 * 1 STSFr. 775.00
- PL 99-120 NMSN, with NTC300 - 14–5...135 °CVetro12×120 mmMemosens® screw plugLog in for availability1 * 1 STSFr. 949.00
- PL A-91-120 pHT VP, with Pt10000 - 14–5...+110 °CVetro12×120 mmVP screw plugLog in for availability1 * 1 STSFr. 622.00
- PL A-91-225 pHT VP, with Pt10000 - 14–5...+110 °CVetro12×225 mmVP screw plugLog in for availability1 * 1 STSFr. 691.00
- PL A-93-120 NMSN, with NTC300 - 14–5...+110 °CVetro12×120 mmMemosens® screw plugLog in for availability1 * 1 STSFr. 722.00
- PL H-91-120 pHT VP, with Pt10000 - 140...+135 °CVetro12×120 mmVP screw plugLog in for availability1 * 1 STSFr. 631.00
- PL H-91-225 pHT VP, with Pt10000 - 140...+135 °CVetro12×225 mmVP screw plugLog in for availability1 * 1 STSFr. 704.00
- PL H-93-120 NMSN, with NTC300 - 140...+135 °CVetro12×120 mmMemosens® screw plugLog in for availability1 * 1 STSFr. 754.00
- PL H-93-225 NMSN, with NTC300 - 140...+135 °CVetro12×225 mmMemosens® screw plugLog in for availability1 * 1 STSFr. 852.00
- PL H-93-325 NMSN, with NTC300 - 140...+135 °CVetro12×325 mmMemosens® screw plugLog in for availability1 * 1 STSFr. 899.00
- PL H-93-360 NMSN, with NTC300 - 140...+135 °CVetro12×360 mmMemosens® screw plugLog in for availability1 * 1 STSFr. 949.00
- PL PETR-120VP, with Pt1000 + redox0 - 140...+130 °CVetro12×120 mmVP screw plugLog in for availability1 * 1 STSFr. 799.00