Robotic tips for BioTek™ pipetting systems
Filtered, low retention, 150 µl, +, +, BioTek™, 10 rack da 96 pezzi
Codice catalogo 613-1887
Nome del fornitore: Thermo Fisher Scientific
Robotic tips for BioTek™ pipetting systems
Codice catalogo 613-1887
Nome del fornitore: Thermo Fisher Scientific
Vai a
Product Details & Documents
These automation tips for BioTek™ pipetting systems undergo a rigorous 15-point quality control program to ensure consistency thus providing accurate and precise performance achieved by superior tip straightness and a low coefficient of variation.
- Sterile pipette tips are certified free of RNase, DNase, DNA, endotoxin, bioburden and pyrogen
- Lot-tested during production on actual workstation for which they are created
- Low percentage coefficient of variation (%CV)
Specifiche
- Confezione:10 rack da 96 pezzi
- Sterili:+
- Volume:150 µl
- Seq:07
- Stile punta:Filtered, low retention
- Per uso con:BioTek™
- Filtro:+
- Cat. No.:613-1887
- Environmentally Preferable:Produzione a basso impatto
- Certificazione ambientale:IRECS; ISO 14001 (Certificate Registration No. EMS 752603)