Conductivity cells
Robust conductivity cells for on-line measuring. Cells are either available with POM shaft or with glass shaft for high chemical resistance.
- Screw plug head with thread Pg 13.5
- LF 1101 versions and LF 1051 with platinum plates
- LF 5101 versions and LF 71 T with platinum rings
- HD electrode: Certified pressure and temperature consistency according to DIN EN 10204
Pressure range: 10 bar
: All electrodes (except LF 1101 T HD) supplied with washer 9980/52 and O-ring type 9932/07. LF 1101 T HD is supplied with HD sealing type 9980/51. Connecting cables need to be ordered separately.
Robust conductivity cells for on-line measuring. Cells are either available with POM shaft or with glass shaft for high chemical resistance.
- Screw plug head with thread Pg 13.5
- LF 1101 versions and LF 1051 with platinum plates
- LF 5101 versions and LF 71 T with platinum rings
- HD electrode: Certified pressure and temperature consistency according to DIN EN 10204
Pressure range: 10 bar
: All electrodes (except LF 1101 T HD) supplied with washer 9980/52 and O-ring type 9932/07. LF 1101 T HD is supplied with HD sealing type 9980/51. Connecting cables need to be ordered separately.
Product Family Options
- DescrizioneTipoConduttanzaCostante di cellaTemperaturaMaterialeØ×LConnettore
- Cella di conducibilità a 2 poliLF 11010 - 200 mS/cm1,0 cm-1−30...+100 °CVetro12×120 mmTesta a spina avvitabile1 * 1 STSFr. 799.00
- Cella di conducibilità a 2 poli, con sensore di temperatura Pt 1000LF 1101 T0 - 200 mS/cm1,0 cm-1−30...+100 °CVetro12×120 mmTesta a spina avvitabile1 * 1 STSFr. 1’180.00
- Cella di conducibilità a 2 poliLF 51010 - 200 mS/cm1,0 cm-10...+80 °CPOM12×120 mmTesta a spina avvitabile1 * 1 STSFr. 640.00
- Cella di conducibilità a 2 poli, con sensore di temperatura Pt 1000LF 5101 T0 - 200 mS/cm1,0 cm-10...+80 °CPOM12×120 mmTesta a spina avvitabile1 * 1 STSFr. 827.00
- 4-pole conductivity cell, with PT1000 temperature sensorLF 71T1X0 - 200 mS/cm1,0 cm-1−30...+100 °CVetro12×120 mmfixed cable, open end1 * 1 STSFr. 1’420.00