Aghi per anestesia spinale, Pencil Point, BD™ Whitacre
BDAM405104
- Codice colore:porpora
- Gauge:24 G
- Tipo:Sterile
- Descrizione:Spinal needle
- Lunghezza:90 mm
- Seq:03
- Conf:25
The BD® Whitacre needles are manufactured with proprietary needle tipping, side-port and low-glare hub technologies.
- Better post-operative patient experience with BD® Whitacre spinal needles that minimise the risk of postdural puncture headache (PDPH)
- Enhanced precision with BD® Whitacre spinal needles that enable visualisation of CSF, aiding proper needle placement
- BD® Whitacre spinal needles yield a distinct tactile 'click' or 'pop' as the pencil-point penetrates the dura, alerting the clinician of proper placement
- Precision-formed side hole helps directional flow of anaesthetic agents and helps reduce the possibility of straddling the dura
- The BD® Whitacre Spinal Needle is supported by 25+ independant published studies