Supporto per provette
Katalognr. 241-0262
Supplier: USBECK Laborgeräte
Specifications
- Lunghezza:130 mm
- Conf:1
- Capacità:30 mm
Specifications
About this item
Test tube holder with finger grip, made from stainless steel.
Test tube holder with finger grip, made from stainless steel.