Orders LinkContactUsLinkComponent
3,4,5-Trimethoxybenzaldehyde
3,4,5-Trimethoxybenzaldehyde
Glass bottle for solids, 100 g
Catalog # MOLE42154644-100G
Marketsource
Supplier:   Molekula
3,4,5-Trimethoxybenzaldehyde
Catalog # MOLE42154644-100G
Supplier:  Molekula
Marketsource
CAS Number:  86-81-7
3,4,5-Trimethoxybenzaldehyde
Glass bottle for solids, 100 g
CAS Number:  86-81-7
Price
Inquire For Price
Log in for availability
1 * 100 g
Product Details & Documents
  • CAS Number:86-81-7
  • Formula:C₁₀H₁₂O₄
  • MDL Number:MFCD00003364
  • Molecular Weight:196.2 g/mol
  • Boiling Pt:311 °C (1013 hPa)
  • Melting Pt:72…75 °C
  • Density:1,367 g/cm³ (20 °C)
  • Storage Temperature:Ambiente
  • EINECS:201-701-6
Specifications
Specifications
  • Tipo di conf.:
    Glass bottle for solids
  • Conf:
    100 g
Specifications