Veratraldehyde
Glass bottle for solids, 500 g
MOLEM43971872
Marketsource
: Molekula
: 3,4-Dimethoxybenzaldehyde
- :120-14-9
- :C₉H₁₀O₃
- :MFCD00003363
- :166.18 g/mol
- :12,10084
- :281 °C (1013 hPa)
- :40…43 °C
- :>100 °C
- :Ambiente
- :204-373-2
- Tipo di conf.:Glass bottle for solids
- Conf:500 g