Penicillin flask, DURAN®
4000 ml, 50 mm
Codice catalogo 391-0370
Nome del fornitore: DWK Life Sciences
Vai a
Product Details & Documents
This penicillin flask is made from clear DURAN® borosilicate glass 3.3.
- Neck is bent upwards which prevents the liquid culture medium from leaking
- Due to the flat, large floor surface, the layer cover of microorganisms can grow evenly high
- The large liquid surface ensures better aeration and thus faster growth of the microorganisms
Typically suitable for preparation of cultures in nutrient media.
Specifiche
- Capacità:4000 ml
- Ø esterno collo:50 mm
- Cat. No.:391-0370