Pipette bulb, with adapter
Catalog # KART200
Supplier: KARTELL
Specifications
- Colore:rosso
- Conf:1
Specifications
About this item
Moulded pipette bulb made of synthetic rubber.
- Three glass ball valves
- Can be controlled by finger pressure
- No metal parts to corrode
- Can be used with all liquids except those that attack rubber
- Use on bulb or cylindrical pipettes of different capacities
Delivery: Supplied with adapter for pipettes with capacities more than 20 ml.