Taccuini da laboratorio per camera bianca, TexWrite® 22
Katalognr. 818-0247
Supplier: Texwipe
Specifications
- Formato carta:216×279 mm
- Tipo:A righe
- Colore:blu
- Descrizione:Blocco di scrittura per laboratorio
- Numero di pagine:96
- Modello:TX5708
- Conf:10
Specifications
About this item
TexWrite® 22 Cleanroom Laboratory Notebooks are the only notebooks available made with synthetic cleanroom bond stationery. Cleanroom stationery is ideal in critical environments where very low particle and fibre generation is important. The pages of the notebook are sewn and bound with a Mylar®-sealed cover. The notebooks are cleaned and double packaged for cleanroom use. An index page is provided and each page is individually numbered for data retrieval ease. All pages contain dual signature lines and the pages are not removable. These features provide security to support Good Laboratory Practices (GLP) for documentation in critical laboratory and manufacturing environments.
- Chemically resistant cover
- Sewn, permanent bound book with dual signature and date lines on each page.
Format: appr. A4.
Colour: Blue.