Screw caps for narrow neck bottles, square, with thread, for bottles "SafeGrip" series
Catalog # KAUT805-85941
Supplier: Kautex
Specifications
- Filettatura:32 mm
- Colore:blu
- Descrizione:Screw closure, with PTFE insert and tamper evident ring
- Materiale:PP
- Conf:1
- Per bottiglie:50 - 500 ml