Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-Esene
1-Esene
1-Esene
1-Esene
EHERC14202100
:
EHRENSTORFER
:
1-Esene
EHERC14202100
:
EHRENSTORFER
:
Conf:
250 mg
Seq:
2590
:
:
84.16 g/mol
:
63 °C (1013 hPa)
:
–140 °C
:
0.675 g/cm³ (20 °C)
:
–26 °C
:
Ambiente
:
209-753-1
:
2370
:
3,II
:
GHS02
:
GHS08
:
592-41-6
:
C₆H₁₂
:
MFCD00009505