Orders LinkContactUsLinkComponent
Cell scrapers and cell spatula
Cell scrapers and cell spatula
Nome del fornitore:   TPP
Cell scrapers and cell spatula
Nome del fornitore:  TPP
Certificates
Cell scrapers and cell spatula
Cell scrapers and cell spatula
5 varianti
Product Details & Documents
Designed for gentle removal and harvesting of tissue cultures in all types of flasks, dishes and tubes.
  • Easier and more effective collection
  • Scraper head made from flexible PE
  • Pivoting scraper head for easy manoeuvering
  • Choice of three different blade widths
  • Small raised knobs on handle for secure grip
  • Lint-free blister packaging
  • User friendly dispenser case
The cell scraper features a pivoting head, which allows changes in blade angle while directly over the culture surface, giving accessibility to adherent cells even in difficult to reach flask corners. The cell lifter has a specially formed blade with sharp edges. It is mainly used with tissue culture flasks with reclosable lid or peel-off foil, or with large tissue culture dishes.
The Triple Bagged (3-B) packaging system meets strict cleanroom standards designed to prevent contamination entering highly sensitive areas of production. The innermost bag 1 is labeled with the product details, that is product number, product description, lot number. Each product is packaged individually in a tear-open bag. Each bag is labeled with a security number: 3 = external, 2 = moderate, 1 = innermost bag. The safety-style peel-open bags are increasingly sized, so that during handling of one bag the next inner bag is not accidentally contaminated.
With the standard packaging, contamination adhering to the outside of the bag could be unintentionally carried into clean room areas. The 3-B packaging system prevents contamination from reaching highly sensitive production areas, thus significantly increasing safety for handling substances in a sterile environment.
Certifications: ISO 9001:2008 certified.
Packaging: Peel-open paper/plastic blister of one.
Product Family Options
Standard packaging
Informazioni sul prodotto
  • Lunghezza totale
    Disponibilità
    Price
  • TPPA99002
    Cell scraper, rotating blade width 13 mm, for 25/75 cm² flasks, 10 cm² tubes, or dishes Ø 40/60 mm, PE
    Cell scraper, rotating blade width 13 mm, for 25/75 cm² flasks, 10 cm² tubes, or dishes Ø 40/60 mm, PE
    240 mm
    Log in per disponibilità
    Log in per disponibilità
    1 * 150 ST
    1 * 150 ST - SFr. 360.00
    SFr. 360.00
    1 * 150 ST
       SFr. 360.00
  • TPPA99003
    Cell scraper, rotating blade width 20 mm, for 75/150 cm² flasks or dishes Ø 100/150 mm, PE
    Cell scraper, rotating blade width 20 mm, for 75/150 cm² flasks or dishes Ø 100/150 mm, PE
    300 mm
    Log in per disponibilità
    Log in per disponibilità
    1 * 150 ST
    1 * 150 ST - SFr. 407.00
    SFr. 407.00
    1 * 150 ST
       SFr. 407.00
  • TPPA99004
    Cell scraper, rotating blade width 25 mm, for 300 cm² flasks, PE
    Cell scraper, rotating blade width 25 mm, for 300 cm² flasks, PE
    362 mm
    Log in per disponibilità
    Log in per disponibilità
    1 * 100 ST
    1 * 100 ST - SFr. 334.00
    SFr. 334.00
    1 * 100 ST
       SFr. 334.00
  • TPPA99010
    Cell lifter, fixed blade width 14 mm, PP
    Cell lifter, fixed blade width 14 mm, PP
    195 mm
    Log in per disponibilità
    Log in per disponibilità
    1 * 150 ST
    1 * 150 ST - SFr. 307.00
    SFr. 307.00
    1 * 150 ST
       SFr. 307.00
Triple bagged packaging for cleanroom appilcations
Informazioni sul prodotto
  • Lunghezza totale
    Disponibilità
    Price
  • TPPA399002
    Cell scraper, rotating blade width 13 mm, for 25/75 cm² flasks, 10 cm² tubes, or dishes Ø 40/60 mm, PE
    Cell scraper, rotating blade width 13 mm, for 25/75 cm² flasks, 10 cm² tubes, or dishes Ø 40/60 mm, PE
    240 mm
    Log in per disponibilità
    Log in per disponibilità
    1 * 40 ST
    1 * 40 ST - SFr. 256.00
    SFr. 256.00
    1 * 40 ST
       SFr. 256.00