Orders LinkContactUsLinkComponent
Glicerina
Glicerina
Marketsource
Supplier:   GERBU BIOCHEMICALS
Glicerina
Supplier:  GERBU BIOCHEMICALS
Marketsource
Certificates
Glicerina
2 Options Available Below
Product Details & Documents
Synonyms:  1,2,3-Propanetriol, Propane-1,2,3-triol, Glycerol
  • CAS Number:56-81-5
  • Formula:HOCH₂CH(OH)CH₂OH
  • MDL Number:MFCD00004722
  • Molecular Weight:92.09 g/mol
  • Merck Index:13,04497
  • Boiling Pt:290 °C (1013 hPa)
  • Melting Pt:18,6 °C
  • Density:1,26 g/cm³ (20 °C)
  • Flash Pt:177 °C
  • Storage Temperature:Ambiente
  • EINECS:200-289-5
Product Family Options
Product Information
  • Tipo di conf.
    Conf
    Availability
    Price
  • bottiglia in vetro
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L
    Specifications:
    Marketsource
    Marketsource
  • 6 L
    Log in for availability
    Log in for availability
    1 * 6 L
    1 * 6 L
    Specifications:
    Marketsource