Orders LinkContactUsLinkComponent
1-Butanolo
1-Butanolo
bottiglia in vetro, 1 L
Catalog # ICNA19400191
Flammable
Corrosive
Irritant
Supplier:   MP Biomedicals
1-Butanolo
Catalog # ICNA19400191
Supplier:  MP Biomedicals
CAS Number:  71-36-3
1-Butanolo
1-Butanolo
bottiglia in vetro, 1 L
CAS Number:  71-36-3
Price
SFr. 126.00
Log in for availability
1 * 1 L
Product Details & Documents
Synonyms:  1-Butyl alcohol, Butyl Alcohol, 1-Hydroxy butane, Propyl carbinol, n-Butanol, n-Butyl Alcohol
  • CAS Number:71-36-3
  • Formula:H₃C(CH₂)₂CH₂OH
  • MDL Number:MFCD00002964
  • Molecular Weight:74.12 g/mol
  • Merck Index:13,01539
  • Boiling Pt:117,6 °C (1013 hPa)
  • Melting Pt:–89 °C
  • Density:0,812 g/cm³ (20 °C)
  • Flash Pt:30 °C
  • Storage Temperature:Ambiente
  • EINECS:200-751-6
  • UN:1120
  • ADR:3,III
  • REACH:01-2119484630-38
Specifications
Specifications
  • Tipo di conf.:
    bottiglia in vetro
  • Conf:
    1 L
Specifications