Hexylamine per sintesi, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: 1-Hexylamine, 1-Aminohexane
CAS Number:111-26-2
:
Formula:H₃C(CH₂)₄CH₂NH₂
MDL Number:MFCD00008240
Boiling Pt:131
Melting Pt:–19 °C
Density:0.766 g/cm³ (20 °C)
Molecular Weight:101.19 g/mol
Flash Pt:27
Storage Temperature:Ambiente
UN:2734
ADR:8 (3) I
:M-Clarity™ quality level = MQ200
EINECS:203-851-8
:GHS02
:GHS06
:GHS05
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Solubility:14 g/l (20 °C)
- Melting Point:-19 °C
- Molar Mass:101.19 g/mol
- Boiling Point:130 - 132 °C (1013 hPa)
- Vapor Pressure:10.6 hPa (20 °C)
- Flash Point:27 °C
- Refractive Index:1.418 (20 °C, 589 nm)
- Explosion Limit:2.1 - 9.3 %(V)
- Density:0.76 g/cm3 (20 °C)
- pH:11.6 (10 g/l, H2O, 20 °C)
- Ignition Point:270 °C
- Viscosity kinematic:1.07 mm2/s (23 °C)
- Synonyms:1-Aminohexane
Specification Test Results
Related Information
Frequently Bought Together