Orders LinkContactUsLinkComponent
4-Methoxybenzonitrile
4-Methoxybenzonitrile
Irritant
Marketsource
Supplier:   COMBI-BLOCKS
4-Methoxybenzonitrile
Supplier:  COMBI-BLOCKS
Marketsource
Certificates
4-Methoxybenzonitrile
3 Options Available Below
Product Details & Documents
Synonyms:  4-Cyanoanisol, Anisonitrile, p-Cyanoanisol, p-Methoxybenzonitrile
  • CAS Number:874-90-8
  • Formula:C₈H₇NO
  • MDL Number:MFCD00001818
  • Molecular Weight:133.15 g/mol
  • Boiling Pt:256 °C (1013 hPa)
  • Melting Pt:57…60 °C
  • Storage Temperature:Ambiente
  • EINECS:212-871-6
  • UN:3439
  • ADR:6.1,III
Product Family Options
Product Information
  • Tipo di conf.
    Conf
    Availability
    Price
  • Bottiglia di vetro
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g
    Specifications:
    Marketsource
    Marketsource
  • Bottiglia di vetro
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g
    Specifications:
    Marketsource
    Marketsource
  • Bottiglia di vetro
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g
    Specifications:
    Marketsource