Giare, con coperchio
25 ml, 47 mm, 32 mm
Codice catalogo 216-3185
Nome del fornitore: Brand
Vai a
Product Details & Documents
PS, with LDPE lid.
- Conical shape
- Good chemical resistance
- Lightweight
Ideal for collecting, storing and transporting samples.
Specifiche
- Ø est:47 mm
- Capacità:25 ml
- Altezza:32 mm