Hydrometer cylinder, Nalgene®
500 ml
Codice catalogo NALG6230-0500
Nome del fornitore: Thermo Fisher Scientific
Hydrometer cylinder, Nalgene®
Codice catalogo NALG6230-0500
Nome del fornitore: Thermo Fisher Scientific
Vai a
Product Details & Documents
Made of PMP, clear.
- Superior chemical resistance
- With broad base
- Good temperature resistance
PMP protect against breakage with the hydrometer cylinder. Lighter in weight than glass. This transparent, autoclavable cylinder can also be used as a lactometer cylinder.
Specifiche
- Capacità:500 ml
- Cat. No.:NALG6230-0500
- Environmentally Preferable:Produzione a basso impatto
- Certificazione ambientale:IRECS; ISO 14001 (Certificate Registration No. EMS 752603)