ContactUsLinkComponent
Sodio diidrogeno fosfato monoidrato
Sodio diidrogeno fosfato monoidrato
  EURLEMR306500
 :  EUROCLONE
 :  
undefined
Sodio diidrogeno fosfato monoidrato
  EURLEMR306500
 :  EUROCLONE
 :  
  • Conf:
    1 KIT
  • Seq:
    0075
  • Environmentally Preferable:
 : Sodium phosphate monobasic monohydrate, Sodium dihydrogen orthophosphate monohydrate, Monosodium phosphate monohydrate, Sodium dihydrogenorthophosphate monohydrate, Sodium orthophosphate monobasic monohydrate
 :
 :137.99 g/mol
 :100
 :2.04 g/cm³ (20 °C)
 :Ambiente
 :231-449-2
 :10049-21-5
 :NaH₂PO₄·H₂O
 :MFCD00149208
 :13,08734