Pipette box holders, Nalgene®
Nome del fornitore: Thermo Fisher Scientific
Pipette box holders, Nalgene®
Nome del fornitore: Thermo Fisher Scientific
Certificates
Informazioni su questo articolo
The acrylic pipette box holder (two compartments) and the angled pipette holder (four compartments) accommodate bags, boxes, pipette canisters or 1- to 100- ml individually-wrapped serological pipettes.
- Compartments sized to fit one box of pipettes (14,6 cm wide opening)
- Storage area for rubber bulbs
- Divider can be removed to form one large storage area
- Can be mounted under the shelf with two brackets
- Angled pipette holder for easy access; prevents pipettes from sliding out
1 varianti
Product Details & Documents
The acrylic pipette box holder (two compartments) and the angled pipette holder (four compartments) accommodate bags, boxes, pipette canisters or 1- to 100- ml individually-wrapped serological pipettes.
- Compartments sized to fit one box of pipettes (14,6 cm wide opening)
- Storage area for rubber bulbs
- Divider can be removed to form one large storage area
- Can be mounted under the shelf with two brackets
- Angled pipette holder for easy access; prevents pipettes from sliding out
Specifications for Product Family
Specifiche
- Catalog No:
- NALG5810-0002
- NALG5810-0004
- Environmentally Preferable:
- Produzione a basso impatto
- Certificazione ambientale:
- NAR Certificate of Retirement
Specifiche
Product Family Options
Informazioni sul prodotto
- ColoreL×P×ADisponibilitàPrice
- NALG5810-0002Portapipette, 2 scompartiPortapipette, 2 scompartitrasparente152×381×152 mmDiscontinued-Specifiche:
More
Specifications
Cat. No.NALG5810-0002Environmentally PreferableCertificazione ambientale - NALG5810-0004Portapipette, 4 scomparti, angolatoPortapipette, 4 scomparti, angolatotrasparente89×292×416 mmLog in per disponibilità1 * 1 STSFr. 827.00Specifiche:
Produzione a basso impatto More
Specifications
Cat. No.NALG5810-0004Environmentally PreferableProduzione a basso impattoCertificazione ambientaleNAR Certificate of RetirementProduzione a basso impatto