OmniTrays, Nunclon™Δ, with enhanced rectangular area
OmniTray, enhanced rectangular area, trasparente, 84 cm², 90 ml
Catalog # 734-2402
Supplier: Thermo Fisher Scientific
OmniTrays, Nunclon™Δ, with enhanced rectangular area
Catalog # 734-2402
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
PS, sterile. Optimised for computerised imaging and spotting.
Specifications
- Colore:trasparente
- Descrizione:OmniTray, enhanced rectangular area
- Superficie coltura:84 cm²
- Volume totale:90 ml
- Cat. No.:734-2402
- Environmentally Preferable:Produzione a basso impatto
- Certificazione ambientale:NAR Certificate of Retirement