Orders LinkContactUsLinkComponent
Pipette tips, gel loading, Corning®
Pipette tips, gel loading, Corning®
Supplier:   Corning
Pipette tips, gel loading, Corning®
Supplier:  Corning
Certificates
Pipette tips, gel loading, Corning®
Pipette tips, gel loading, Corning®
4 Options Available Below
Product Details & Documents
These gel-loading pipette tips feature a capillary end that allows easy access into vertical and horizontal electrophoresis gels.
  • Disposable
  • Certified RNase-/DNase-free and non pyrogenic
Ordering information: Available in flat and round tip configurations. Flat tips are designed for use in 0,2 and 0,4 mm sequencing gels. Round tips are designed for use in SDS-PAGE 0,5 mm gels.
Documents
Product Family Options
Product Information
  • Stile punta
    Volume
    Sterili
    Lunghezza
    Colore puntale
    Confezione
    Availability
    Price
  • Micro, flat (capillary thickness: 0,2 mm)
    0,2 - 10 µl
    -
    83 mm
    Trasparente
    2 racks, 200 each
    Log in for availability
    Log in for availability
    1 * 400 ST
    1 * 400 ST - SFr. 220.00
    SFr. 220.00
    1 * 400 ST
       SFr. 220.00
  • Round (capillary thickness: 0,5 mm)
    1 - 200 µl
    -
    83 mm
    Trasparente
    2 racks, 200 each
    Log in for availability
    Log in for availability
    1 * 400 ST
    1 * 400 ST - SFr. 144.00
    SFr. 144.00
    1 * 400 ST
       SFr. 144.00
  • Flat (capillary thickness: 0,4 mm)
    1 - 200 µl
    -
    83 mm
    Trasparente
    2 racks, 200 each
    Log in for availability
    Log in for availability
    1 * 400 ST
    1 * 400 ST - SFr. 206.00
    SFr. 206.00
    1 * 400 ST
       SFr. 206.00
  • Flat (capillary thickness: 0,2 mm)
    1 - 200 µl
    -
    83 mm
    Trasparente
    2 racks, 200 each
    Log in for availability
    Log in for availability
    1 * 400 ST
    1 * 400 ST - SFr. 257.00
    SFr. 257.00
    1 * 400 ST
       SFr. 257.00