Cappucci per cleanroom, Kimtech™ A5
Supplier: KIMBERLY CLARK
Certificates
About this item
Il cappuccio regolabile è in tessuto traspirante SMS che lo rende confortevole come un qualsiasi altro tessuto. Elastico traforato e cuciture interne.
- More breathable than many other leading brands
- Average bacterial filtration efficiency (BFE) of 96%
- Pull-down ties in the back to assist in the gowning process
- No gaps between hood and mask ensures exhaled air is filtered
- Vacuum packed for added sterility assurance
Adatto per Cleanroom Classe 100, M 3,5, ISO 5.
Certifications: Type 6 [PB]; Cat. III
Packaging: Triple-bagged.
Specifications
- Catalog No:
- 113-7732
- 114-9642
- Environmentally Preferable:
- Riduzione degli sprechi
- Classe di cleanroom:
- 100/ISO 5
- 100/ISO 5
- Applicazioni per abiti:
- Cleanroom
- Cleanroom
Specifications
Related Information
Frequently Bought Together