Orders LinkContactUsLinkComponent
Iodio | Avantor
Iodio
Health
Irritant
Environment
Marketsource
Supplier:   Roth Carl
Iodio
Supplier:  Roth Carl
Marketsource
Certificates
3 Options Available Below
Product Details & Documents
  • CAS Number:7553-56-2
  • Formula:I₂
  • MDL Number:MFCD00011355
  • Molecular Weight:253.81 g/mol
  • Merck Index:13,05036
  • Boiling Pt:184,3 °C (1013 hPa)
  • Melting Pt:114 °C
  • Density:4,9 g/cm³ (20 °C)
  • Storage Temperature:Ambiente
  • EINECS:231-442-4
  • UN:3495
  • ADR:8,III
Product Family Options
Product Information
  • Tipo di conf.
    Conf
    Availability
    Price
  • Bottiglia di vetro
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - SFr. 104.30
    SFr. 104.30
    1 * 100 g
       SFr. 104.30
    Specifications:
    Marketsource
    Marketsource
  • Bottiglia di vetro
    250 g
    Log in for availability
    Log in for availability
    1 * 250 g
    1 * 250 g - SFr. 190.80
    SFr. 190.80
    1 * 250 g
       SFr. 190.80
    Specifications:
    Marketsource
    Marketsource
  • Bottiglia di vetro
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg - SFr. 666.00
    SFr. 666.00
    1 * 1 kg
       SFr. 666.00
    Specifications:
    Marketsource
    Marketsource