Piastre per 96 pozzetti per attivazione di cellule T Corning® BioCoat™
Katalognr. 734-1075
Supplier: Corning
Specifications
- Confezione:1/busta
- Descrizione:Piastre per analisi a 96 pozzetti per attivazione di cellule T, anti-CD3 umanizzati
- Environmentally Preferable:Produzione a basso impatto
- Certificazione ambientale:ISO 14001 - Bedford
- Conf:5
Specifications
About this item
Corning BioCoat T-Cell activation plates are precoated with high quality BD Pharmingen™ CD3 antibodies. Available for use with mouse or human T-cells, Corning BioCoat T-Cell activation plates offer lot-to-lot consistency and come individually packaged with lids for ease of use.
- Tested for ability to proliferate mouse splenocytes or human PBMCs
- Tested and found negative for bacteria and fungi
Applications include:
- T-Cell activation
- Cytokine production
- Cytokine mRNA quantitation
- Co-stimulation
- Studies of drug effects on T-cell function