Modelli animali e cellule vegetali
Cell structure model, 160×150×90 mm
Codice catalogo 761-0014
Marketsource
Nome del fornitore: 3B Scientific
Modelli animali e cellule vegetali
Codice catalogo 761-0014
Nome del fornitore: 3B Scientific
Marketsource
Vai a
Product Details & Documents
Modelli ingranditi di cellula animale e cellula vegetale.
- 12 electron microscopic illustrations of different cell structures
- Structures are numbered and identified
- Reproducible illustrations for use in testing
Enable visual teaching about their structures, as well as their similarities, and differences.
Informazioni per l'ordine: Supplied with teacher's aid in English.
Specifiche
- Descrizione:Cell structure model
- L×P×A:160×150×90 mm