Measuring magnifiers
Informazioni su questo articolo
These technical magnifiers can be used for every standard application.
- Achromatic lens made of high quality silicate for a distortion-free image with pinpoint clarity
- Fine focusing via easy-grip focusing ring
- Housing made from glass fibre reinforced polyamide
There are different precision scales available for use with the magnifiers.
Bestellinformationen: Supplied with standard crosshair scale (no. 115201), measuring range 20 mm, 1/10 mm graduation.
2 varianti
Product Details & Documents
These technical magnifiers can be used for every standard application.
- Achromatic lens made of high quality silicate for a distortion-free image with pinpoint clarity
- Fine focusing via easy-grip focusing ring
- Housing made from glass fibre reinforced polyamide
There are different precision scales available for use with the magnifiers.
Bestellinformationen: Supplied with standard crosshair scale (no. 115201), measuring range 20 mm, 1/10 mm graduation.
Product Family Options
Informazioni sul prodotto
Accessories
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Crosshair scale (Ø = 23 mm)
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Multi scale (Ø = 23 mm)
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Metric hole scale (Ø = 23 mm)
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Straight line scale (Ø = 23 mm)
Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.
Andernfalls erhalten Sie allgemeine Empfehlungen..