FlashGel™ DNA system
Informazioni su questo articolo
FlashGel™ DNA system offers a rapid method for separating DNA and watching DNA migration as it happens. The system consists of enclosed, disposable, precast agarose FlashGe™ DNA cassettes and an electrophoresis unit with built in transilluminator (FlashGel™ dock) that provides both separation and detection.
- Separates DNA fragments 10 bp to 4 kb in 2 to 7 minutes
- Gel runs can be monitored at the bench, in real time without UV light
- At least five times more sensitive than gels stained with ethidium bromide
Certifications: Some components and technology of the FlashGel™ system are sold under licensing agreements. The nucleic acid stain in this product is manufactured and sold under license from Molecular Probes, Inc for use only in research applications or quality control, and is covered by pending and issued patents. The FlashGel™ Dock technology is licensed from Clare Chemical Research, Inc. The electrophoresis technology is licensed from Temple University.
Informazioni per l'ordine: Supplied as a complete system (including FlashGel™ dock), separate components or as a starter kit (without FlashGel™ camera). The markers and dye are in a ready to load format, optimised for use with the system and recommended for best performance. The starter kit contains 9× FlashGel™ DNA cassettes (1,2%, 12+1 well); 1 ml FlashGel™ loading dye; and 150 µl FlashGel™ DNA marker (100 bp to 4 Kb).
Attenzione: Gel cannot be accessed. Unsuitable for recovery.
7 varianti
Product Details & Documents
FlashGel™ DNA system offers a rapid method for separating DNA and watching DNA migration as it happens. The system consists of enclosed, disposable, precast agarose FlashGe™ DNA cassettes and an electrophoresis unit with built in transilluminator (FlashGel™ dock) that provides both separation and detection.
- Separates DNA fragments 10 bp to 4 kb in 2 to 7 minutes
- Gel runs can be monitored at the bench, in real time without UV light
- At least five times more sensitive than gels stained with ethidium bromide
Certifications: Some components and technology of the FlashGel™ system are sold under licensing agreements. The nucleic acid stain in this product is manufactured and sold under license from Molecular Probes, Inc for use only in research applications or quality control, and is covered by pending and issued patents. The FlashGel™ Dock technology is licensed from Clare Chemical Research, Inc. The electrophoresis technology is licensed from Temple University.
Informazioni per l'ordine: Supplied as a complete system (including FlashGel™ dock), separate components or as a starter kit (without FlashGel™ camera). The markers and dye are in a ready to load format, optimised for use with the system and recommended for best performance. The starter kit contains 9× FlashGel™ DNA cassettes (1,2%, 12+1 well); 1 ml FlashGel™ loading dye; and 150 µl FlashGel™ DNA marker (100 bp to 4 Kb).
Attenzione: Gel cannot be accessed. Unsuitable for recovery.
Documents
Certificates
Cerca per numero di lottoSpecifications for Product Family
Specifiche
- Catalog No:
- LONZ57067
- 733-1610
- 733-1613
- 733-1611
- 733-1614
- 733-1615
- LONZ57051
- Modello:
- FlashGel™ DNA
- FlashGel™ DNA
- FlashGel™ DNA
- FlashGel™ DNA
- FlashGel™ DNA
- FlashGel™ DNA
- FlashGel™ DNA
- Dimensione gel largh.×lungh.:
- 84×70 mm
- 84×70 mm
- 84×70 mm
- 84×70 mm
- 84×70 mm
- 84×70 mm
- 84×70 mm
- Range di separazione (bp):
- 1,2% agarose: 50 - 4000
2,2% agarose: 10 - 1000 - 1,2% agarose: 50 - 4000
2,2% agarose: 10 - 1000 - 1,2% agarose: 50 - 4000
2,2% agarose: 10 - 1000 - 1,2% agarose: 50 - 4000
2,2% agarose: 10 - 1000 - 1,2% agarose: 50 - 4000
2,2% agarose: 10 - 1000 - 1,2% agarose: 50 - 4000
2,2% agarose: 10 - 1000 - 1,2% agarose: 50 - 4000
2,2% agarose: 10 - 1000
- Stoccaggio (cassette):
- Room temperature for up to 5 months from manufacture
- Room temperature for up to 5 months from manufacture
- Room temperature for up to 5 months from manufacture
- Room temperature for up to 5 months from manufacture
- Room temperature for up to 5 months from manufacture
- Room temperature for up to 5 months from manufacture
- Room temperature for up to 5 months from manufacture
- Volume del pozzetto:
- 2 - 5 µl
- 2 - 5 µl
- 2 - 5 µl
- 2 - 5 µl
- 2 - 5 µl
- 2 - 5 µl
- 2 - 5 µl
- Dimensione cassetta L×P×A:
- 107×115×17 mm
- 107×115×17 mm
- 107×115×17 mm
- 107×115×17 mm
- 107×115×17 mm
- 107×115×17 mm
- 107×115×17 mm
- Dimensione stazione L×P×A:
- 120×134×54 mm
- 120×134×54 mm
- 120×134×54 mm
- 120×134×54 mm
- 120×134×54 mm
- 120×134×54 mm
- 120×134×54 mm
Specifiche
Product Family Options
Informazioni sul prodotto
- DisponibilitàPrice
- LONZ57067FlashGel™ system, including FlashGel™ dock, FlashGel™ camera, 9× FlashGel™ DNA cassettes (1,2%, 12+1-well format), FlashGel™ loading dye and FlashGel™ DNA markerFlashGel™ system, including FlashGel™ dock, FlashGel™ camera, 9× FlashGel™ DNA cassettes (1,2%, 12+1-well format), FlashGel™ loading dye and FlashGel™ DNA markerLog in per disponibilità1 * 1 STSFr. 3’900.00Specifiche:
More
Specifications
Cat. No.LONZ57067ModelloFlashGel™ DNADimensione gel largh.×lungh.84×70 mmRange di separazione (bp)1,2% agarose: 50 - 4000
2,2% agarose: 10 - 1000Stoccaggio (cassette)Room temperature for up to 5 months from manufactureVolume del pozzetto2 - 5 µlDimensione cassetta L×P×A107×115×17 mmDimensione stazione L×P×A120×134×54 mm - 733-1610FlashGel™ system for DNA, starter kit (without camera)FlashGel™ system for DNA, starter kit (without camera)Log in per disponibilità1 * 1 KITSFr. 1’810.00Specifiche:
More
Specifications
Cat. No.733-1610ModelloFlashGel™ DNADimensione gel largh.×lungh.84×70 mmRange di separazione (bp)1,2% agarose: 50 - 4000
2,2% agarose: 10 - 1000Stoccaggio (cassette)Room temperature for up to 5 months from manufactureVolume del pozzetto2 - 5 µlDimensione cassetta L×P×A107×115×17 mmDimensione stazione L×P×A120×134×54 mm - 733-1613FlashGel™ DNA cassettes, 1,2% agarose, 16+1 double tierFlashGel™ DNA cassettes, 1,2% agarose, 16+1 double tierLog in per disponibilità1 * 9 STSFr. 346.00Specifiche:
More
Specifications
Cat. No.733-1613ModelloFlashGel™ DNADimensione gel largh.×lungh.84×70 mmRange di separazione (bp)1,2% agarose: 50 - 4000
2,2% agarose: 10 - 1000Stoccaggio (cassette)Room temperature for up to 5 months from manufactureVolume del pozzetto2 - 5 µlDimensione cassetta L×P×A107×115×17 mmDimensione stazione L×P×A120×134×54 mm - 733-1611FlashGel™ DNA cassettes, 1,2% agarose, 12+1 single tierFlashGel™ DNA cassettes, 1,2% agarose, 12+1 single tierLog in per disponibilità1 * 9 STSFr. 346.00Specifiche:
More
Specifications
Cat. No.733-1611ModelloFlashGel™ DNADimensione gel largh.×lungh.84×70 mmRange di separazione (bp)1,2% agarose: 50 - 4000
2,2% agarose: 10 - 1000Stoccaggio (cassette)Room temperature for up to 5 months from manufactureVolume del pozzetto2 - 5 µlDimensione cassetta L×P×A107×115×17 mmDimensione stazione L×P×A120×134×54 mm - 733-1614FlashGel™ DNA cassettes, 2,2% agarose, 12+1 single tierFlashGel™ DNA cassettes, 2,2% agarose, 12+1 single tierLog in per disponibilità1 * 9 STSFr. 346.00Specifiche:
More
Specifications
Cat. No.733-1614ModelloFlashGel™ DNADimensione gel largh.×lungh.84×70 mmRange di separazione (bp)1,2% agarose: 50 - 4000
2,2% agarose: 10 - 1000Stoccaggio (cassette)Room temperature for up to 5 months from manufactureVolume del pozzetto2 - 5 µlDimensione cassetta L×P×A107×115×17 mmDimensione stazione L×P×A120×134×54 mm - 733-1615FlashGel™ DNA cassettes, 2,2% agarose, 16+1 double tierFlashGel™ DNA cassettes, 2,2% agarose, 16+1 double tierLog in per disponibilità1 * 9 STSFr. 346.00Specifiche:
More
Specifications
Cat. No.733-1615ModelloFlashGel™ DNADimensione gel largh.×lungh.84×70 mmRange di separazione (bp)1,2% agarose: 50 - 4000
2,2% agarose: 10 - 1000Stoccaggio (cassette)Room temperature for up to 5 months from manufactureVolume del pozzetto2 - 5 µlDimensione cassetta L×P×A107×115×17 mmDimensione stazione L×P×A120×134×54 mm - LONZ57051FlashGel™ recovery cassettes, 1,2% agarose, 8+1 double tierFlashGel™ recovery cassettes, 1,2% agarose, 8+1 double tierLog in per disponibilità1 * 9 STSFr. 346.00Specifiche:
More
Specifications
Cat. No.LONZ57051ModelloFlashGel™ DNADimensione gel largh.×lungh.84×70 mmRange di separazione (bp)1,2% agarose: 50 - 4000
2,2% agarose: 10 - 1000Stoccaggio (cassette)Room temperature for up to 5 months from manufactureVolume del pozzetto2 - 5 µlDimensione cassetta L×P×A107×115×17 mmDimensione stazione L×P×A120×134×54 mm
Accessories
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Formaldehyde sample buffer
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- RNA marker (0,5 - 9 kb), 50 µL
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- FlashGel™ recovery kit includes recovery cassettes 1,2%, 8+1 double-tier well, loading dye, recovery buffer, QuantLadder, visualisation glasses, and maskFlashGel™ recovery kit includes recovery cassettes 1,2%, 8+1 double-tier well, loading dye, recovery buffer, QuantLadder, visualisation glasses, and maskLONZ57064Log in per disponibilità1 * 1 KIT - SFr. 1’050.00
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- FlashGel™ DNA marker (50 bp to 1 Kb), for 2,2% cassettesFlashGel™ DNA marker (50 bp to 1 Kb), for 2,2% cassettes733-1616Log in per disponibilità1 * 500 µl - SFr. 456.00
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Marcatore RNA (da 0,5 bp a 9 Kb)
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- FlashGel™ DNA QuantLadder (100 bp to 1500 bp), 50 applicationsFlashGel™ DNA QuantLadder (100 bp to 1500 bp), 50 applications733-1621Log in per disponibilità1 * 250 µl - SFr. 456.00
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- FlashGel™ DNA marker (100 bp to 4 Kb), for 1.2% cassettesFlashGel™ DNA marker (100 bp to 4 Kb), for 1.2% cassettes733-1620Log in per disponibilità1 * 500 µl - SFr. 456.00
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- FlashGel™ DNA marker (100 bp to 3 Kb), for 2 tier cassettesFlashGel™ DNA marker (100 bp to 3 Kb), for 2 tier cassettes733-1622Log in per disponibilità1 * 500 µl - SFr. 456.00
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- FlashGel™ loading dye, 5X, RNA native sample buffer (contains xylene cyanol)FlashGel™ loading dye, 5X, RNA native sample buffer (contains xylene cyanol)733-1619Log in per disponibilità1 * 5 mL - SFr. 363.00
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- FlashGel™ dock
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- FlashGel™ recovery buffer, 2× 500 μl
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- FlashGel™ power supply
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- FlashGel™ power supply pack, includes FlashGel™ dockFlashGel™ power supply pack, includes FlashGel™ dockLONZ57069Log in per disponibilità1 * 1 KIT - SFr. 2’620.00