Extra-wide mini horizontal electrophoresis systems, Owl™ D3-14 and A6
Codice catalogo 730-0109
Nome del fornitore: THERMO OWL SCIENTIFIC
Specifiche
- Modello:Owl D3-14
- Descrizione:Minisistema orizzontale su gel extralargo
- Volume tampone:800 ml
- Dimensione gel largh.×lungh.:230×140 mm
- Unità Largh. x Lungh. x P:300×270×110 mm
- Conf:1
Specifiche
Informazioni su questo articolo
The Owl™ D3-14 and A6 wide gel systems offer a simple, convenient and rapid means of screening multiple samples on a single agarose gel. Ideal for screening PCR samples, plasmid preps, restriction mapping and cloning.
- 25 to 200 samples can be run simultaneously on one gel, producing clear tight banding patterns with no ‘smiling'
- Sample loading is greatly enhanced using microwell format combs engineered specifically for use with the system
- Samples may be loaded directly from a 96-well plate, 8 or 12 at a time when using microwell combs and a multichannel pipette, speeding up transfer and reducing the risk of sample order error
Delivery: Each system includes buffer chamber, SuperSafe lid with attached power leads, gasketed UVT gel tray, external caster and four combs (Owl D3-14: 50-microwell, 1,5 mm thick, Owl A6: 2×25- and 2×50-microwell, 1,5 mm thick).