Orders LinkContactUsLinkComponent
Microwave digestion vessels
Microwave digestion vessels
Nome del fornitore:   PARR
Microwave digestion vessels
Nome del fornitore:  PARR
Certificates
Microwave digestion vessels
Microwave digestion vessels
4 varianti
Product Details & Documents
Digestion vessels with PTFE liners designed specifically for the microwave. A high-strength, transparent, microwaveable material surrounds the chemically inert PTFE liners, allowing these vessels to be placed directly in a microwave oven for specific, rapid heating. The high temperatures and pressures developing in the vessels as a result of this special procedure result in an extremely aggressive digestion process, without overheating the outer sheath. With form-fitting seals and integrated overpressure protection. Special ovens are not required; the vessels can be used in any lab or household microwave.
Product Family Options
Informazioni sul prodotto
  • Disponibilità
    Price
  • 458-4510
    Microwave digestion vessel, Model 4781, 23 ml
    Microwave digestion vessel, Model 4781, 23 ml
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 Un.
    1 * 1 Un. - SFr. 4’070.00
    SFr. 4’070.00
    1 * 1 Un.
       SFr. 4’070.00
    Specifiche:
    More
    Specifications
    Cat. No.
    458-4510
  • 458-4511
    PTFE cup and cover with O-ring, 23 ml
    PTFE cup and cover with O-ring, 23 ml
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 Un.
    1 * 1 Un. - SFr. 565.00
    SFr. 565.00
    1 * 1 Un.
       SFr. 565.00
    Specifiche:
    More
    Specifications
    Cat. No.
    458-4511
  • 458-4520
    Microwave digestion vessel, Model 4782, 45 ml
    Microwave digestion vessel, Model 4782, 45 ml
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 Un.
    1 * 1 Un.
    Specifiche:
    More
    Specifications
    Cat. No.
    458-4520
  • 458-4521
    PTFE cup and cover with O-ring, 45 ml
    PTFE cup and cover with O-ring, 45 ml
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 Un.
    1 * 1 Un. - SFr. 799.00
    SFr. 799.00
    1 * 1 Un.
       SFr. 799.00
    Specifiche:
    More
    Specifications
    Cat. No.
    458-4521