Orders LinkContactUsLinkComponent
Nessler cylinders
Nessler cylinders
Nessler cylinders
  • Optically polished, flat base
  • With pouring spout
Product Family Options
Forma alta
  • Descrizione
  • Nessler cylinders, 50 ml, tall form, Ø×H: 21×265, 50 ml division
    1 * 1 ST
    1 * 1 ST - SFr. 63.30
    SFr. 63.30
    1 * 1 ST
       SFr. 63.30
  • Nessler cylinders, 100 ml, tall form, Ø×H: 26×320, 50 + 100 ml division
    1 * 1 ST
    1 * 1 ST - SFr. 74.60
    SFr. 74.60
    1 * 1 ST
       SFr. 74.60
  • Nessler cylinders, 100 ml, tall form, Ø×H: 26×320, 100 ml division
    1 * 1 ST
    1 * 1 ST - SFr. 69.40
    SFr. 69.40
    1 * 1 ST
       SFr. 69.40
Forma bassa
  • Descrizione
  • Nessler cylinders, 50 ml, low form, Ø×H: 26×175, 50 ml division
    1 * 1 ST
    1 * 1 ST - SFr. 57.50
    SFr. 57.50
    1 * 1 ST
       SFr. 57.50
  • Nessler cylinders, 100 ml, low form, Ø×H: 32×220, 50 + 100 ml division
    1 * 1 ST
    1 * 1 ST - SFr. 82.60
    SFr. 82.60
    1 * 1 ST
       SFr. 82.60