Certificate of calibration
Codice catalogo 620-1494
Nome del fornitore: DICKSON
Specifiche
- Descrizione:Calibrazione a un punto NIST Traceable®
- Conf:1
- Per:Chart recorders
Specifiche
Informazioni su questo articolo
A NIST certificate provides that your Dickson instrument had been properly calibrated and is traceable back to the National Institute of Standards and Technology (NIST).
- Includes documentation to one Dickson pre-selected point after calibration
- Allows you to comply with audits
- Helps keep track of instrument calibration requirements
- Easy recalibration ordering information also included
This type of documented calibration will satisfy most quality and compliance situations. This one-point NIST offers proof of the accuracy of Dickson''s quality instruments.