Tripod stand for flow cups
Stativo per piastra (260 mm)
Catalog # 769-1150
Supplier: Industrial Physics
Jump to:
Product Details & Documents
The spirit level is easy to balance thanks to the adjustable feet of the unit.
- Stainless steel ring and PVC legs
- Tripod stand with built-in spirit level
Suitable for all types of viscosity cups.
Documents
TQC Company Video
Videos
Specifications
- Descrizione:Stativo per piastra (260 mm)
- Seq:920