Scintillation vials, WHEATON®
20,00 ml, 27×57 mm, Yes
Codice catalogo 548-0704
Nome del fornitore: DWK Life Sciences
Vai a
Product Details & Documents
These clear scintillation vials with screw cap are made from soda-lime glass.
- Low background count
- Fitted with foil lined urea cap
Specifiche
- Tappo incluso:Yes
- Ø×A:27×57 mm
- Capacità:20,00 ml